Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSC Monoclonal Antibody (4C5D5)
GREENER_CHOICE

Catalog No. PIMA531756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531756 Supplier Invitrogen™ Supplier No. MA531756
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31756 has been tested in indirect ELISA.

This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSC
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4C5D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GSC
Gene Accession No. P56915
Gene Alias GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS
Gene Symbols GSC
Host Species Mouse
Immunogen Purified recombinant fragment of human GSC (AA: 191-257) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.