Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541494 Supplier Invitrogen™ Supplier No. PA541494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LEALFVQNQY PDVSTRERLA GRIRLREERV EVWFKNRRAK WRHQKRASAS Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 85%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%; Zebrafish: 91%.

Goosecoidlike (GSCL) resides in the critical region for VCFS/DGS on 22q11. Velocardiofacial syndrome (VCFS) is phenotypically related to DiGeorge syndrome (DGS) and both syndromes are associated with hemizygous 22q11 deletions. Because many of the tissues and structures affected in VCFS/DGS derive from the pharyngeal arches of the developing embryo, it is believed that haploinsufficiency of a gene involved in embryonic development may be responsible for its etiology.Goosecoidlike (GSCL), a homeodomain-containing gene, resides in the critical region for VCFS/DGS on 22q11. Velocardiofacial syndrome (VCFS) is a developmental disorder characterized by conotruncal heart defects, craniofacial anomalies, and learning disabilities. VCFS is phenotypically related to DiGeorge syndrome (DGS) and both syndromes are associated with hemizygous 22q11 deletions. Because many of the tissues and structures affected in VCFS/DGS derive from the pharyngeal arches of the developing embryo, it is believed that haploinsufficiency of a gene involved in embryonic development may be responsible for its etiology. The gene is expressed in a limited number of adult tissues, as well as in early human development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSC2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GSC2
Gene Accession No. O15499
Gene Alias 4930568H22Rik; goosecoid homebox 2; goosecoid homeobox 2; GSC2; GSC-2; GSCL; GSC-L; homeobox protein goosecoid-2; Homeobox protein goosecoid-like; LOW QUALITY PROTEIN: homeobox protein goosecoid-2
Gene Symbols GSC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GSCL (aa 141-190).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2928
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.