Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSDMA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554757
Description
Immunogen sequence: VGALTELSEA QQKLLVKSME KKILPVQLKL VESTMEQNFL LDKEGVFPLQ PELLSSLGDE ELTLTEALVG LSGLEVQRSG PQYMWDPDTL PRLCALYAGL SLLQQLTKAS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
GSDMA induces apoptosis.
Specifications
GSDMA | |
Polyclonal | |
Unconjugated | |
GSDMA | |
BB149167; FKSG9; gasdermin 1; gasdermin A; gasdermin A1; gasdermin-1; Gasdermin-A; gasdermin-A1; Gsdm; GSDM1; Gsdma; Gsdma1; H312E | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284110 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96QA5 | |
GSDMA | |
Recombinant protein corresponding to Human GSDMA. Recombinant protein control fragment (Product #RP-93393). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction