Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSDMA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119970
This item is not returnable.
View return policy
Description
Positive Control: Human skin tissue lysate, rat skin tissue lysate. Subcellular Location: Cytosol, perinuclear region; Cell membrane.
GSDMA induces apoptosis.
Specifications
GSDMA | |
Polyclonal | |
Unconjugated | |
GSDMA | |
BB149167; FKSG9; gasdermin 1; gasdermin A; gasdermin A1; gasdermin-1; Gasdermin-A; gasdermin-A1; Gsdm; GSDM1; Gsdma; Gsdma1; H312E | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284110, 360619 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96QA5 | |
GSDMA | |
Recombinant protein within human GSDMA aa 1-200. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction