Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSDMA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524813

Catalog No. PIPA524813


Only null left
Add to Cart

Description

Description

GSDMA Polyclonal Antibody for Western Blot, IHC (P), Flow

GSDMA induces apoptosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GSDMA
Polyclonal
Unconjugated
GSDMA
BB149167; FKSG9; gasdermin 1; gasdermin A; gasdermin A1; gasdermin-1; Gasdermin-A; gasdermin-A1; Gsdm; GSDM1; Gsdma; Gsdma1; H312E
Rabbit
Antigen affinity chromatography, Protein A
RUO
284110, 57911
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q96QA5, Q9EST1
GSDMA
KLH conjugated synthetic peptide between 66-94 amino acids from the N-terminal region of human GSDMA.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.