Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSDMB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562601 Supplier Invitrogen™ Supplier No. PA562601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKSLGSEDSR NMKEKLEDME SVLKDLTEEK RKDVLNSLAK CLGKEDIRQD LEQRVSEVLI SGELHMEDPD KPLLSSLFNA AGVLV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 28%.

This gene encodes a member of the gasdermin-domain containing protein family. Other gasdermin-family genes are implicated in the regulation of apoptosis in epithelial cells, and are linked to cancer. Multiple transcript variants encoding different isoforms have been found for this gene. Additional variants have been described, but they are candidates for nonsense-mediated mRNA decay (NMD) and are unlikely to be protein-coding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSDMB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GSDMB
Gene Accession No. Q8TAX9
Gene Alias gasdermin B; gasdermin-B; Gasdermin-like protein; GSDMB; GSDML; PP4052; PRO2521
Gene Symbols GSDMB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GSDMB. Recombinant protein control fragment (Product #RP-93177).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55876
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.