Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSDMC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524259
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524259 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524259 Supplier Invitrogen™ Supplier No. PA524259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GSDMC Polyclonal Antibody for Western Blot, IHC (P), Flow

The N-terminal moiety promotes pyroptosis. May be acting by homooligomerizing within the membrane and forming pores (PubMed:27281216). The physiological relevance of this observation is unknown (Probable). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSDMC
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GSDMC
Gene Accession No. Q9BYG8
Gene Alias gasdermin C; gasdermin C1; gasdermin-C; Gasdermin-C1; Gsdmc; Gsdmc1; melanoma-derived leucine zipper, extra-nuclear factor; melanoma-derived leucine zipper-containing extranuclear factor; MLZE; mMLZE
Gene Symbols GSDMC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 218-246 amino acids from the central region of human GSDMC.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56169
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.