Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSDMC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555303
Description
Immunogen sequence: LPVGRIEEPF WQNFKHLQEE VFQKIKTLAQ LSKDVQDVMF YSILAMLRDR GALQDLMNML ELDSSGHLDG PGGAILKKLQ QDSNHAWFNP KDP Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 53%.
The N-terminal moiety promotes pyroptosis. May be acting by homooligomerizing within the membrane and forming pores (PubMed:27281216). The physiological relevance of this observation is unknown (Probable). [UniProt].
Specifications
GSDMC | |
Polyclonal | |
Unconjugated | |
GSDMC | |
gasdermin C; gasdermin C1; gasdermin-C; Gasdermin-C1; Gsdmc; Gsdmc1; melanoma-derived leucine zipper, extra-nuclear factor; melanoma-derived leucine zipper-containing extranuclear factor; MLZE; mMLZE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYG8 | |
GSDMC | |
Recombinant protein corresponding to Human GSDMC. Recombinant protein control fragment (Product #RP-93820). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction