Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSDMC Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989809
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9809 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9809 Supplier Bioss Supplier No. BS16332R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The N-terminal moiety promotes pyroptosis. May be acting by homooligomerizing within the membrane and forming pores. The physiological relevance of this observation is unknown.

Specifications

Antigen GSDMC
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene GSDMC
Gene Accession No. Q9BYG8
Gene Alias gasdermin C; gasdermin C1; Gasdermin-C; Gasdermin-C1; Gsdmc; Gsdmc1; melanoma-derived leucine zipper, extra-nuclear factor; Melanoma-derived leucine zipper-containing extranuclear factor; Mlze; mMLZE
Gene Symbols GSDMC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GSDMC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56169
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.