Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSDMD Monoclonal Antibody (A8D7)
GREENER_CHOICE

Catalog No. FEMA550595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550595 Supplier Invitrogen™ Supplier No. MA550595
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: PC-3 cell lysate, NIH/3T3 cell lysate, THP-1 cell lysate. Predicted band size: 53 kDa Subcellular Location: Cytosol, Inflammasome; Secreted, Cell membrane.

GSDMD (gasdermin-D) promotes pyroptosis in response to microbial infection and danger signals. It is produced by the cleavage of gasdermin-D by inflammatory caspases CASP1 or CASP4 in response to canonical, as well as, non-canonical (such as cytosolic LPS) inflammasome activators. After cleavage, GSDMD moves to the plasma membrane where it strongly binds to inner leaflet lipids including phosphatidylinositol 4-phosphate, phosphatidylinositol (4,5)-biphosphate, phosphatidylinositol (3,4,5)-biphosphate, phosphatidic acid, and phosphatidylserine. GSDMD also exhibits bactericidal activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSDMD
Applications Western Blot
Classification Monoclonal
Clone A8D7
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene GSDMD
Gene Accession No. P57764, Q9D8T2
Gene Alias 1810036L03Rik; AW558049; DF5L; Dfna5l; FKSG10; gasdermin D; gasdermin domain containing 1; gasdermin domain-containing protein 1; Gasdermin-D; Gasdermin-D, C-terminal; Gasdermin-D, N-terminal; GSDMD; GSDMDC1; GSDMD-CT; GSDMD-NT; M2-4
Gene Symbols GSDMD
Host Species Mouse
Immunogen Recombinant protein within human GSDMD aa 101-300/484.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69146, 79792
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.