Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSDMD Monoclonal Antibody (A8D8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA550596
Description
Positive control: PC-3 cell lysate, SiHa cell lysate, THP-1 cell lysate. Predicted band size: 53 kDa Subcellular Location: Cytosol, Inflammasome; Secreted, Cell membrane.
GSDMD (gasdermin-D) promotes pyroptosis in response to microbial infection and danger signals. It is produced by the cleavage of gasdermin-D by inflammatory caspases CASP1 or CASP4 in response to canonical, as well as, non-canonical (such as cytosolic LPS) inflammasome activators. After cleavage, GSDMD moves to the plasma membrane where it strongly binds to inner leaflet lipids including phosphatidylinositol 4-phosphate, phosphatidylinositol (4,5)-biphosphate, phosphatidylinositol (3,4,5)-biphosphate, phosphatidic acid, and phosphatidylserine. GSDMD also exhibits bactericidal activity.
Specifications
GSDMD | |
Monoclonal | |
2 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P57764 | |
GSDMD | |
Recombinant protein within human GSDMD aa 101-300/484. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
A8D8 | |
Unconjugated | |
GSDMD | |
1810036L03Rik; AW558049; DF5L; Dfna5l; FKSG10; gasdermin D; gasdermin domain containing 1; gasdermin domain-containing protein 1; Gasdermin-D; Gasdermin-D, C-terminal; Gasdermin-D, N-terminal; GSDMD; GSDMDC1; GSDMD-CT; GSDMD-NT; M2-4 | |
Mouse | |
Protein A | |
RUO | |
79792 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction