Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSDMD Mouse anti-Human, Clone: 2E6C7, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 663871IG150UL

Description
FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.Specifications
GSDMD | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P57764 | |
GSDMD | |
GSDMD Fusion Protein Ag16827 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Western Blot | |
2E6C7 | |
Unconjugated | |
GSDMD | |
DF5L, DFNA5L, FKSG10, gasdermin D, GSDMD, GSDMDC1 | |
Mouse | |
Protein A | |
RUO | |
79792 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction