Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSDMD Recombinant Rabbit Monoclonal Antibody (8X4F4), Invitrogen™

Catalog No. FEMA551265
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551265 Supplier Thermo Scientific Supplier No. MA551265

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DGPAGAVLEC LVLSSGMLVP ELAIPVVYLL GALTMLSETQ HKLLAEALES QTLLGPLELV GSLLEQSAPW QERSTMSLPP GLLGNSWGEG APAWVLLDEC GLELGEDTPH VCWEPQAQGR MCALYASLAL LSGLSQEPH

GSDMD Recombinant Monoclonal antibody specifically detects GSDMD in Human, Mouse, Rat samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSDMD
Applications Immunocytochemistry
Classification Recombinant Monoclonal
Clone 8X4F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene GSDMD
Gene Accession No. P57764, Q9D8T2
Gene Alias 1810036L03Rik, AW558049, DF5L, Dfna5l, FKSG10, gasdermin D, gasdermin domain containing 1, gasdermin domain-containing protein 1, Gasdermin-D, Gasdermin-D, C-terminal, Gasdermin-D, N-terminal, GSDMD, GSDMDC1, GSDMD-CT, GSDMD-NT, M2-4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 346-484 of human GSDMD (NP_079012.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315084, 69146, 79792
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.