Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140067
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140067 Supplier Invitrogen™ Supplier No. PA5140067
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VSLSEPATQQ ASLDVEKPVG VAASLSDIPK AAEPGKLEQV RPQELSRVQE LAPASGEKAR LSEAPGGKKS LSMLHYIRG Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 55%.

This gene encodes a proline-rich protein with coiled coil domains that may be a subunit of a BRAF35-HDAC (BHC) histone deacetylase complex. This gene may function as an oncogene in breast cancer and enhanced expression of the encoded protein has been observed in breast cancer patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GSE1
Gene Accession No. Q14687
Gene Alias 2210013I18Rik; CRHSP24; CTC-786C10.1; fj87f12; genetic suppressor element 1; GSE1; Gse1 coiled-coil protein; Gse1 coiled-coil protein homolog; Kiaa0182; mKIAA0182; RGD1562686; si:dkey-265o13.4; wu:fj87f12
Gene Symbols GSE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GSE1. Recombinant protein control fragment (Product #RP-108836).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23199
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.