Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100588 Supplier Invitrogen™ Supplier No. PA5100588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KIAA0182.

This gene encodes a proline-rich protein with coiled coil domains that may be a subunit of a BRAF35-HDAC (BHC) histone deacetylase complex. This gene may function as an oncogene in breast cancer and enhanced expression of the encoded protein has been observed in breast cancer patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GSE1
Gene Accession No. Q14687, Q3U3C9
Gene Alias 2210013I18Rik; CRHSP24; CTC-786C10.1; fj87f12; genetic suppressor element 1; GSE1; Gse1 coiled-coil protein; Gse1 coiled-coil protein homolog; Kiaa0182; mKIAA0182; RGD1562686; si:dkey-265o13.4; wu:fj87f12
Gene Symbols GSE1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GSE1(Accession Q14687), corresponding to amino acid residues L239-L289.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23199, 307913, 382034
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.