Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114024
Description
Immunogen sequence: VLEFKCKHSK SFKENPNCLP HHHQCFPRRL SSAAPTVGPL TSYHQYHNQP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
May cause the redistribution of PAPOLB from the cytosol to the endoplasmic reticulum.
Specifications
GSG1 | |
Polyclonal | |
Unconjugated | |
GSG1 | |
germ cell associated 1; germ cell-specific gene 1 protein; GSG1; testicular secretory protein Li 20; UNQ709/PRO1360 | |
Rabbit | |
Affinity chromatography | |
RUO | |
83445 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2KHT4 | |
GSG1 | |
Synthetic peptide directed towards the C terminal region of human GSG1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction