Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSG1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107137 Supplier Invitrogen™ Supplier No. PA5107137
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 93%.

As a component of the inner core of AMPAR complex, modifies AMPA receptor (AMPAR) gating.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSG1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene GSG1L
Gene Accession No. Q6UXU4
Gene Alias AI852434; C230098I05Rik; g1l; G630023A01Rik; germ cell-specific gene 1-like protein; Gm637; GSG1 like; GSG1L; GSG1-like; GSG1-like protein; KTSR5831; PRO19651; RGD1562278; UNQ5831/PRO19651
Gene Symbols GSG1L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human GSG1L (aa 100-150).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146395
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.