Learn More
GSG1L Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 173281AP150UL
Description
This antibody can recognize all the 4 isoforms of GSG1L.
As a component of the inner core of AMPAR complex, modifies AMPA receptor (AMPAR) gating.Specifications
| GSG1L | |
| Polyclonal | |
| Unconjugated | |
| GSG1L | |
| GSG1 like, GSG1 like protein, GSG1L, PRO19651 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146395 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6UXU4 | |
| GSG1L | |
| GSG1L Fusion Protein Ag11307 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.