Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103339 Supplier Invitrogen™ Supplier No. PA5103339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GSG2.

GSG2 (haspin) is a conserved kinase required for mitosis. GSG2 phosphorylates histone H3 and is viewed as a potential therapeutic target for oncology. GSG2 is required for normal alignment of chromosomes at metaphase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSG2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HASPIN
Gene Accession No. Q8TF76
Gene Alias germ cell associated 2 (haspin); germ cell associated 2, haspin; germ cell-specific gene 2 protein; GSG2; haploid germ cell-specific nuclear protein kinase; HASPIN; H-haspin; Serine/threonine-protein kinase haspin
Gene Symbols HASPIN
Host Species Rabbit
Immunogen A synthesized peptide derived from human GSG2(Accession Q8TF76), corresponding to amino acid residues Q38-P88.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83903
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.