Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 1016790A
GREENER_CHOICE

Catalog No. 643310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-331-0 10 mg
64-335-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-331-0 Supplier Tocris Bioscience™ Supplier No. 6433/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent TRPV4 agonist

GSK 1016790A is a potent TRPV4 agonist; elicits Ca2+ influx in human and mouse TRPV4-expressing HEK cells (EC50 values of 2.1 and 18 nM, respectively). Enhances insulin mRNA expression. Increases ERK1/2 phosphorylation and NO production. Exhibits no activity at TRPM8 and TRPA1 (20 μM) channels. Induces bladder overactivity in TRPV4+/+ mice.

Specifications

Quantity 10 mg
Molecular Formula C28H32Cl2N4O6S2
Formula Weight 655.61
Product Type Agonist
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.