Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 1702934A
GREENER_CHOICESDP

Catalog No. 650850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-085-0 50 mg
65-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-085-0 Supplier Tocris Bioscience™ Supplier No. 6508/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective TRPC3/6 activator

GSK 1702934A is a potent and selective TRPC3/6 activator (EC50 = 80 and 440 nM for TRPC3 and 6, respectively). Exhibits no activity at TRPV4, TRPA1, M1, M4, CaV1.2, hERG, NaV1.5, or CXCR5 receptors at concentrations <10 μmol/L. Induces TRPC3/6-currents in HEK293 cells transduced with recombinant human TRPC3/6. Directly activates TRPC channels through by-passing phospholipase-C signaling. Also induces calcium signals in HEK-TRPC6 cells (pEC50 = 6.6).

Specifications

Chemical Name or Material 1,3-Dihydro-1-[1-[(5,6,7,8-tetrahydro-4H-cyclohepta[b]thien-2-yl)carbonyl]-4-piperidinyl]-2H-benzimidazol-2-one
CAS 924377-85-5
Quantity 50 mg
Molecular Formula C22H25N3O2S
Formula Weight 395.52
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.