Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 1838705
GREENER_CHOICE

Catalog No. 511110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-111-0 10 mg
51-115-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-111-0 Supplier Tocris Bioscience™ Supplier No. 5111/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent IR and IGF1R inhibitor; also inhibits anaplastic lymphoma kinase (ALK)

GSK 1838705 is a potent insulin receptor (IR) and insulin-like growth factor-1 receptor (IGF1R) inhibitor (IC50 values are 1.6 and 2 nM, respectively). Also inhibits anaplastic lymphoma kinase (ALK) (IC50 = 0.5 nM). Displays > 800-fold selectivity for IR, IGFR1 and ALK over a panel of 44 kinases including JNK. Blocks proliferation of cancer cell lines in vitro, and causes complete regression of ALK-dependent tumors in vivo. Orally bioavailable.

Specifications

Quantity 10 mg
Product Type GSK 1838705
Purity >98%
Molecular Weight (g/mol) 532.57
Inhibitors GSK 1838705

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.