Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 2033
GREENER_CHOICE

Catalog No. 569410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-941-0 10 mg
56-945-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-941-0 Supplier Tocris Bioscience™ Supplier No. 5694/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent LXR antagonist

GSK 2033 is a potent LXR antagonist (pIC50 = 7.5). Enhances T-cell proliferation and blocks T 0901317 -antiproliferative activity on T-cells. Cell permeable.

Specifications

Chemical Name or Material 2,4,6-Trimethyl-N-[[3′-(methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl]-N-[[5-(trifluoromethyl)-2-furanyl]methyl]benzenesulfonamide
CAS 1221277-90-2
Quantity 10 mg
Target LXR-like Receptor Antagonists
Molecular Formula C29H28F3NO5S
Formula Weight 591.66
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.