Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GSK 2239633A
GREENER_CHOICESDP

Catalog No. 707150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-715-0 50 mg
70-711-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-715-0 Supplier R&D Systems™ Supplier No. 7071/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent CCR4 antagonist

GSK 2239633A is a potent CCR4 antagonist (pIC50 = 7.83 in a 35S-GTPγS competition assay). Inhibits thymus activation-regulated chemokine (TARC) -induced F-actin polymerization in human whole blood.

Specifications

Chemical Name or Material N-[[3-[[3-[[(5-Chloro-2-thienyl)sulfonyl]amino]-4-methoxy-1H-indazol-1-yl]methyl]phenyl]methyl]-2-hydroxy-2-methylpropanamide
CAS 1240516-71-5
Quantity 50 mg
Product Line Tocris
Molecular Formula C24H25ClN4O5S2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.