Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 2256098
GREENER_CHOICESDP

Catalog No. 778610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-861-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 77-861-0 Supplier Tocris Bioscience™ Supplier No. 7786/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent focal adhesion kinase (FAK) inhibitor

GSK 2256098 is a potent, reversible and ATP-competitive focal adhesion kinase (FAK) inhibitor (IC50 values are 8.5 nM, 12 nM and 15 nM in U87MG, A549 and OVCAR8 cell lines respectively). Inhibits Y397 phosphorylation and motility, growth and survival of pancreatic ductal adenocarcinoma cells.

Specifications

Chemical Name or Material 2-[[5-Chloro-2-[[3-methyl-1-(1-methylethyl)-1H-pyrazol-5-yl]amino]-4-pyridinyl]amino]-N-methoxybenzamide
CAS 1224887-10-8
Quantity 10 mg
Target Focal Adhesion Kinase Inhibitors
Molecular Formula C20H23ClN6O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.