Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 2879552
SDP

Catalog No. 812850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-285-0 50 mg
81-281-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-285-0 Supplier Tocris Bioscience™ Supplier No. 8128/50
Add to Cart
Add to Cart

Lysine-Specific Demethylase 1 (LSD1) inhibitor

GSK 2879552 is a selective lysine-specific demethylase 1 (LSD1) inhibitor. When combined with other small molecules (EPZ-5676, N-methyl-d-aspartate and Bay K 8644), GSK 2879552 accelerates maturation of human pluripotent stem cell-derived neurons. GSK 2879552 inhibits the growth of small cell lung carcinoma (SCLC) and AML lines. In vivo, GSK 2879552 (1.5 mg/kg, p.o.) demonstrates tumor growth inhibition in SCLC xenograph mouse models.

Specifications

Chemical Name or Material 4-[[4-[[[(1R,2S)-2-Phenylcyclopropyl]amino]methyl]-1-piperidinyl]methyl]benzoic acid dihydrochloride
CAS 1902123-72-1
Quantity 50 mg
Target Histone Demethylase Inhibitors
Molecular Formula C23H28N2O2.2HCl
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.