Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK-3 alpha Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297994AF405
Description
GSK-3 alpha Polyclonal specifically detects GSK-3 alpha in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GSK-3 alpha | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK-3 alpha | |
Produced in rabbits immunized with E. coli-derived Human GSK-3 alpha fragment. | |
0.1 mL | |
Cell Biology, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
2931 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction