Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK-3 beta Antibody (272536), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2506X
Description
GSK-3 beta Monoclonal antibody specifically detects GSK-3 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
GSK-3 beta | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
272536 | |
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
E. coli-derived recombinant human GSK-3 beta , Met1-Thr420, Accession # P49841 | |
0.1 mL | |
Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
2932 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction