Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3 beta Antibody (3D10) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-4980567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147470SS 0.025 mL
NBP147470 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP147470SS Supplier Novus Biologicals Supplier No. NBP147470SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GSK-3 beta
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3D10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Simple Western 1:50, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Gene Accession No. P49841
Gene Alias EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform
Gene Symbols GSK3B
Host Species Mouse
Immunogen Purified recombinant fragment of human GSK3 beta expressed in E. coli. [UniProt# P49841]
Molecular Weight of Antigen 46 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2932
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.