Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK-3 beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $487.50
Specifications
Antigen | GSK-3 beta |
---|---|
Applications | Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GSK-3 beta Polyclonal specifically detects GSK-3 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated.Specifications
GSK-3 beta | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
A synthetic peptide corresponding to a sequence within amino acids 300 to the C-terminus of human GSK-3 beta (NP_001139628.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown | |
Unconjugated | |
Rabbit | |
Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
PBS with 50% glycerol, pH7.3. | |
2932 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title