Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GSK-3 beta Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB2506SP
This item is not returnable.
View return policy
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human, Mouse, Rat samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
GSK-3 beta | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL, CyTOF-ready | |
P49841 | |
GSK3B | |
E. coli-derived recombinant human GSK-3 beta Met1-Thr420 Accession # P49841 | |
25 μg | |
Primary | |
Detects human, mouse, and rat GSK-3 beta in ELISAs and Western blots. Does not cross-react with recombinant human GSK-3 alpha . | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
272536 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2932 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction