Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GSK 3787
GREENER_CHOICESDP

Catalog No. 396150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-615-0 50 mg
39-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-615-0 Supplier R&D Systems™ Supplier No. 3961/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective PPARδ antagonist

Potent and selective peroxisome proliferator-activated receptor δ (PPARδ) antagonist (pIC50 = 6.6). Displays no measurable affinity for PPARα or PPARγ in vitro (pIC50< 5).

Chemical Identifiers

CAS 188591-46-0
Molecular Formula C15H12ClF3N2O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 392.78
Purity >99%
Molecular Weight (g/mol) 392.78
Chemical Name or Material GSK 3787
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.