Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 4112
GREENER_CHOICE

Catalog No. 366310R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-631-0R 10 mg
36-635-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-631-0R Supplier Tocris Bioscience™ Supplier No. 3663/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Rev-Erbα agonist

GSK 4112 is a selective Rev-Erbα agonist (EC50 = 250 nM). Enhances recruitment of nuclear receptor co-repressor (NCoR) peptide to Rev-Erbα; causes acute suppression of Bmal1 gene transcription. Induces phase shifts in the circadian clock.

Specifications

Quantity 10 mg
Product Type GSK 4112
Purity >98%
Molecular Weight (g/mol) 396.89
Inhibitors GSK 4112

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.