Learn More
Tocris Bioscience™ GSK 429286
Description
Specifications
Specifications
| Purity | >97% |
| Gene Alias | GSK429286A |
| Quantity | 1 mg |
| Product Type | GSK 429286 |
| Molecular Weight (g/mol) | 432.37 |
| Inhibitors | GSK 429286 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.