Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 429286
SDP

Catalog No. 372650R Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-265-0R 50 mg
37-261-R 1 mg
37-261-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 37-265-0R Supplier Tocris Bioscience™ Supplier No. 3726/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective Rho-kinase (ROCK) inhibitor

GSK 429286 is a selective Rho-kinase inhibitor (IC50 values are 14, 780 and 1940 nM for ROCK1, RSK and p70S6K respectively). Reverses adrenaline-induced contraction of the rat aortic ring (IC50 = 190 nM) and causes a dose-dependent decrease in mean arterial blood pressure in spontaneous hypertensive rats. Orally active.

Specifications

Purity >97%
Gene Alias GSK429286A
Quantity 50 mg
Product Type GSK 429286
Molecular Weight (g/mol) 432.37
Inhibitors GSK 429286

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.