Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 583
GREENER_CHOICESDP

Catalog No. 64805 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-805 5 mg
64-802-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-805 Supplier Tocris Bioscience™ Supplier No. 6480/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent RIP2 kinase inhibitor; orally bioavailable and cell permeable

GSK 583 is a potent RIPK2 inhibitor (IC50 values are 2 and 5 nM at rat and human kinases, respectively). Displays selectivity for RIP2K over p38α, VEGFR2 and a panel of 300 kinases. Exhibits some inhibition at BRK and Aurora A. Also displays similar binding affinity, but no detectable functional activity at RIP3. Active in vivo. Moderately orally bioavailable and cell permeable.

Specifications

Chemical Name or Material 6-[(1,1-Dimethylethyl)sulfonyl]-N-(5-fluoro-1H-indazol-3-yl)-4-quinolinamine
CAS 1346547-00-9
Quantity 5 mg
Molecular Formula C20H19FN4O2S
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.