Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 690693
GREENER_CHOICESDP

Catalog No. 414410 Shop All Novus Biologicals Products
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
41-441-0 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-441-0 Supplier Tocris Bioscience™ Supplier No. 4144/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Akt kinase inhibitor. Antitumor

ATP-competitive pan-Akt kinase inhibitor (IC50 values are 2, 13 and 9 nM of Akt1, 2 and 3 respectively). Also exhibits some inhibition for AMPK, PKA and PAK and PKC isoforms (IC50 < 100 nM). Displays antiproliferative and apoptotic effects in tumor cell lines.

Specifications

CAS 937174-76-0
Quantity 10 mg
Molecular Formula C21H27N7O3
Formula Weight 425.48
Purity >98%
Molecular Weight (g/mol) 425.493
Chemical Name or Material GSK 690693
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.