Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK'872
GREENER_CHOICESDP

Catalog No. 649250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-925-0 50 mg
64-921-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-925-0 Supplier Tocris Bioscience™ Supplier No. 6492/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective RIP3 kinase inhibitor

GSK'872 is a potent and selective receptor activating protein kinase 3 (RIP3) inhibitor (IC50 = 1.3 nM). Exhibits >1000-fold selectivity for RIP3 over a panel of 300 other kinases including RIP1. Inhibits TNF-induced cell death in 3T3-SA cells and TLR3-induced necrosis in fibroblasts in vitro. Blocks necroptosis of primary human neutrophils from whole blood. Also induces apoptosis at higher concentrations (3 - 10 μM).

Specifications

Chemical Name or Material N-5-Benzothiazolyl-6-[(1-methylethyl)sulfonyl]-4-quinolinamine
CAS 1346546-69-7
Quantity 50 mg
Molecular Formula C19H17N3O2S2
Formula Weight 383.49
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.