Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK 9027
GREENER_CHOICE

Catalog No. 411610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-161-0 10 mg
41-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-161-0 Supplier Tocris Bioscience™ Supplier No. 4116/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Glucocorticoid receptor agonist

Glucocorticoid receptor agonist (pIC50 = 8). Inhibits production of the proinflammatory mediator IL-6 in vivo.

Chemical Identifiers

CAS 1229096-88-1
Molecular Formula C27H19F4N3O2S

Specifications

Quantity 10 mg
Formula Weight 525.52
Purity >98%
Molecular Weight (g/mol) 525.522
Chemical Name or Material GSK 9027
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.