Learn More
Description
Cell permeable ester derivative, GSK J5 and Active Analog also available.
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 389.45 |
| Physical Form | Powder |
| Color | White |
| Purity | >98% |
| Molecular Weight (g/mol) | 388.451 |
| Chemical Name or Material | GSK J2 |
Safety and Handling
| Recommended Storage | Desiccate at RT |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

