Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK J2
GREENER_CHOICE

Catalog No. 468810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-881-0 10 mg
46-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-881-0 Supplier Tocris Bioscience™ Supplier No. 4688/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inactive isomer of GSK J1 (Cat. No. 4593)

Inactive control of GSK J1 (IC50 > 100 μM for inhibition of JMJD3/UTX).

Cell permeable ester derivative, GSK J5 and Active Analog also available.

Chemical Identifiers

CAS 1394854-52-4
Molecular Formula C22H22N5O2-

Specifications

Quantity 10 mg
Formula Weight 389.45
Physical Form Powder
Color White
Purity >98%
Molecular Weight (g/mol) 388.451
Chemical Name or Material GSK J2
Recommended Storage Desiccate at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.