Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GSK J4
GREENER_CHOICESDP

Catalog No. 459450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-945-0 50 mg
45-941-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-945-0 Supplier R&D Systems™ Supplier No. 4594/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Histone KDM inhibitor; cell permeable

Histone lysine demethylase (KDM) inhibitor; blocks demethylation of histone H3K27. Attenuates lipopolysaccharide (LPS)-induced proinflammatory cytokine production in primary human macrophages (IC50 = 9 μM for the inhibition of TNFα release). Cell permeable. Ethyl ester derivative of GSK J1 .

Negative Control also available.

Chemical Identifiers

CAS 1373423-53-0
Molecular Formula C24H27N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 417.5
Purity >99%
Molecular Weight (g/mol) 417.513
Chemical Name or Material GSK J4
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.