Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK J4
GREENER_CHOICESDP

Catalog No. 459410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-941-0 10 mg
45-945-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-941-0 Supplier Tocris Bioscience™ Supplier No. 4594/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Histone KDM inhibitor; cell permeable

Histone lysine demethylase (KDM) inhibitor; blocks demethylation of histone H3K27. Attenuates lipopolysaccharide (LPS)-induced proinflammatory cytokine production in primary human macrophages (IC50 = 9 μM for the inhibition of TNFα release). Cell permeable. Ethyl ester derivative of GSK J1 .

Negative Control also available.

Chemical Identifiers

CAS 1373423-53-0
Molecular Formula C24H27N5O2

Specifications

Quantity 10 mg
Formula Weight 417.5
Purity >99%
Molecular Weight (g/mol) 417.513
Chemical Name or Material GSK J4
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.