Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK J5, Tocris Bioscience™

Catalog No. 468950
Change view
Click to view available options
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
46-895-0 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-895-0 Supplier R&D Systems Supplier No. 4689/50
This item is not returnable. View return policy

Inactive isomer of GSK J4 (Cat. No. 4594); cell permeable

Inactive isomer of GSK J4 (Cat. No. 4594); also cell permeable ester derivative of the inactive control, GSK J2 (Cat. No. 4688)

Specifications

CAS 1394854-51-3
Color White
Molecular Formula C24H27N5O2
MDL Number MFCD27991275
Quantity 50 mg
Synonym gsk j5, gsk-j5, ethyl 3-2-pyridin-3-yl-6-2,3,4,5-tetrahydro-1h-3-benzazepin-3-yl pyrimidin-4-yl amino propanoate, n-2-3-pyridinyl-6-1,2,4,5-tetrahydro-3h-3-benzazepin-3-yl-4-pyrimidinyl-beta-alanine ethyl ester
InChI Key LQPGVGSKBNXQDU-UHFFFAOYSA-N
SMILES CCOC(=O)CCNC1=NC(=NC(=C1)N1CCC2=CC=CC=C2CC1)C1=CN=CC=C1
IUPAC Name ethyl 3-{[2-(pyridin-3-yl)-6-(2,3,4,5-tetrahydro-1H-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoate
Molecular Weight (g/mol) 417.51
PubChem CID 78243729
Formula Weight 417.5
Percent Purity >98%
Physical Form Powder
Chemical Name or Material GSK J5
Show More Show Less
Recommended Storage Store at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.