Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GSK2578215A
GREENER_CHOICE

Catalog No. 462925
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-292-5 25 mg
46-295 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-292-5 Supplier Tocris Bioscience™ Supplier No. 4629/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective LRRK2 inhibitor; brain penetrant

GSK2578215A is a potent LRRK2 inhibitor (IC50 values are 8.9 and 10.1 nM for LRRK2[G2019S] mutant and wild-type LRRK2 respectively). Displays selectivity for LRRK2 over a panel of 460 other kinases. Blocks Ser910 and Ser935 phosphorylation in vitro and in peripheral tissues in vivo. Brain penetrant.

Specifications

Chemical Name or Material 5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinylbenzamide
CAS 1285515-21-0
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C24H18FN3O2
Formula Weight Observed MW: 399.42
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.