Learn More
Promega GSK3 alpha Kinase Enzyme System, 1mg
Catalog No. PRV3052 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3052 1 mg
1 options
Catalog No. PRV3052 Supplier Promega Supplier No. V3052
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human GSK3 alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 alpha is a multifunctional protein serine kinase implicated in the control of several regulatory proteins.
  • Easily Screen and Profile GSK3α Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg