Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSK3 alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587470 Supplier Invitrogen™ Supplier No. PA587470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGTQLPNNRP LPPLFNFSAG ELSIQPSLNA ILIPPHLRSP AGTTTLTPSS QALTETPTSS DWQSTDATPT LTNSS; Positive Samples: U-87MG, A-431, HeLa, B-cell, Mouse liver.

Glycogen synthase kinase 3 alpha (GSK3A) is a multifunctional protein serine/threonine kinase implicated in the control of several regulatory proteins and transcription factors. (GSK3A Alias: DKFZp686D0638). Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK3 alpha
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gsk3a
Gene Accession No. P49840, Q2NL51
Gene Alias 2700086H06Rik; DKFZp686D0638; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 alpha b; glycogen synthase kinase 3 alpha b; glycogen synthase kinase-3 alpha; glycogen synthase kinase-3 alpha; GSK3; GSK-3 alpha; GSK-3[a]; GSK-3[b]; gsk3a; gsk3ab; gsk-3-alpha; OTTHUMP00000198368; Serine/threonine-protein kinase GSK3A
Gene Symbols Gsk3a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 409-483 of human GSK3alpha (NP_0639372).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2931, 606496
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.