Learn More
Promega GSK3 beta Kinase Enzyme System, 1mg
Catalog No. PRV1992 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV1992 1 mg
1 options
Catalog No. PRV1992 Supplier Promega Supplier No. V1992
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human GSK3 beta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. GSK3 beta is a serine-threonine protein kinase originally identified as the kinase that phosphorylates and inhibits glycogen synthase.
  • Easily Screen and Profile GSK3β Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C