Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSK3B Monoclonal Antibody (3D10)
GREENER_CHOICE

Catalog No. PIMA515597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515597 Supplier Invitrogen™ Supplier No. MA515597
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15597 targets GSK3B in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human, mouse, Non-human primate, and Rat samples. The MA5-15597 immunogen is purified recombinant fragment of human GSK3B expressed in E. Coli.. MA5-15597 detects GSK3B which has a predicted molecular weight of approximately 46kDa.

GSK3B is a proline-directed serine/threonine protein kinase involved in energy metabolism, neuronal cell development and body pattern formation. It is a negative regulator of glucose homeostasis and is involved in energy metabolism, inflammation, ER-stress, mitochondrial dysfunction, and apoptotic pathways. GSK3B is a critical component of the Wnt/beta-catenin signaling pathway, and its activity results in the inhibition of Wnt signaling by mediating the degradation of beta-catenin. GSK3B is also involved in the Hedgehog, Notch, and insulin signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK3B
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3D10
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene GSK3B
Gene Accession No. P18266, P49841, Q9WV60
Gene Alias 7330414F15Rik; 8430431H08Rik; C86142; EC 2.7.11.26; FA; Factor A; fb68h05; fk80d11; glycogen synthase kinase 3; glycogen synthase kinase 3 beta; glycogen synthase kinase 3 beta L homeolog; glycogen synthase kinase 3 beta variant 1; glycogen synthase kinase 3 beta variant 2; glycogen synthase kinase 3 beta variant 3; glycogen synthase kinase 3 beta variant 4; glycogen synthase kinase 3 beta variant 5; glycogen synthase kinase 3 beta, genome duplicate a; glycogen synthase kinase-3 beta; glycogen synthase kinase-3 beta splice variant X1; glycogen synthase kinase-3 beta splice variant X2; glycogen synthase kinase-3 beta splice variant X3; GSK 3 beta; GSK 3B; gsk3; GSK-3; GSK3 beta; GSK-3 beta; GSK-3 beta GSK3B; GSK-3[b]; Gsk3b; GSK-3b; gsk3b.L; gsk3ba; gsk3beta; GSK-3beta; gsk3-beta; GSK3beta isoform; intracellular kinase; pp37; serine/threonine-protein kinase GSK3B; wu:fb68h05; wu:fk80d11; XELAEV_180112401mg; xgsk-3; Xgsk-3 protein
Gene Symbols GSK3B
Host Species Mouse
Immunogen Purified recombinant fragment of human GSK3B expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2932, 56637, 84027
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.