Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSK3B Monoclonal Antibody (ZG004)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 399500

Encompass_Preferred

Catalog No. 39-950-0


Only null left
Add to Cart

Description

Description

GSK3B Monoclonal Antibody for Western Blot, IHC (P), ELISA

GSK3B is a proline-directed serine/threonine protein kinase involved in energy metabolism, neuronal cell development and body pattern formation. It is a negative regulator of glucose homeostasis and is involved in energy metabolism, inflammation, ER-stress, mitochondrial dysfunction, and apoptotic pathways. GSK3B is a critical component of the Wnt/beta-catenin signaling pathway, and its activity results in the inhibition of Wnt signaling by mediating the degradation of beta-catenin. GSK3B is also involved in the Hedgehog, Notch, and insulin signaling pathways.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GSK3B
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide
P18266, P49841, Q9WV60
GSK3B
Human GSK3 beta.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1 κ
ELISA, Immunohistochemistry (Paraffin), Western Blot
ZG004
Unconjugated
GSK3B
7330414F15Rik; 8430431H08Rik; C86142; EC 2.7.11.26; FA; Factor A; fb68h05; fk80d11; glycogen synthase kinase 3; glycogen synthase kinase 3 beta; glycogen synthase kinase 3 beta L homeolog; glycogen synthase kinase 3 beta variant 1; glycogen synthase kinase 3 beta variant 2; glycogen synthase kinase 3 beta variant 3; glycogen synthase kinase 3 beta variant 4; glycogen synthase kinase 3 beta variant 5; glycogen synthase kinase 3 beta, genome duplicate a; glycogen synthase kinase-3 beta; glycogen synthase kinase-3 beta splice variant X1; glycogen synthase kinase-3 beta splice variant X2; glycogen synthase kinase-3 beta splice variant X3; GSK 3 beta; GSK 3B; gsk3; GSK-3; GSK3 beta; GSK-3 beta; GSK-3 beta GSK3B; GSK-3[b]; Gsk3b; GSK-3b; gsk3b.L; gsk3ba; gsk3beta; GSK-3beta; gsk3-beta; GSK3beta isoform; intracellular kinase; pp37; serine/threonine-protein kinase GSK3B; wu:fb68h05; wu:fk80d11; XELAEV_180112401mg; xgsk-3; Xgsk-3 protein
Mouse
Affinity chromatography
RUO
2932, 56637, 84027
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.